Learn More
Abnova™ Human QARS Partial ORF (NP_005042, 677 a.a. - 775 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005042 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5859 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16143985
|
Abnova™
H00005859-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16133985
|
Abnova™
H00005859-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. In metazoans, 9 aminoacyl-tRNA synthetases specific for glutamine (gln), glutamic acid (glu), and 7 other amino acids are associated within a multienzyme complex. Although present in eukaryotes, glutaminyl-tRNA synthetase (QARS) is absent from many prokaryotes, mitochondria, and chloroplasts, in which Gln-tRNA(Gln) is formed by transamidation of the misacylated Glu-tRNA(Gln). Glutaminyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family. [provided by RefSeq]
Sequence: FIHWVSQPLMCEVRLYERLFQHKNPEDPTEVPGGFLSDLNLASLHVVDAALVDCSVALAKPFDKFQFERLGYFSVDPDSHQGKLVFNRTVTLKEDPGKVSpecifications
NP_005042 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLNRS/PRO2195 | |
QARS | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5859 | |
QARS (Human) Recombinant Protein (Q01) | |
FIHWVSQPLMCEVRLYERLFQHKNPEDPTEVPGGFLSDLNLASLHVVDAALVDCSVALAKPFDKFQFERLGYFSVDPDSHQGKLVFNRTVTLKEDPGKV | |
RUO | |
QARS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |