missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RAB11FIP1 Full-length ORF (NP_001002233.1, 1 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001002233.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80223 |
Molecular Weight (g/mol) | 91.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144853
|
Abnova™
H00080223-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134853
|
Abnova™
H00080223-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Proteins of the large Rab GTPase family (see RAB1A; MIM 179508) have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP1 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 [PubMed 11495908]).[supplied by OMIM]
Sequence: MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKMSpecifications
NP_001002233.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKM | |
RUO | |
RAB11FIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
80223 | |
RAB11FIP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E2214/FLJ22524/FLJ22622/MGC78448/NOEL1A/RCP/rab11-FIP1 | |
RAB11FIP1 | |
Recombinant | |
wheat germ expression system |