missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAB11FIP1 Full-length ORF (NP_001002233.1, 1 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080223-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134853

  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Proteins of the large Rab GTPase family (see RAB1A; MIM 179508) have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP1 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 [PubMed 11495908]).[supplied by OMIM]

Sequence: MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKM
Specifications

Specifications

NP_001002233.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686E2214/FLJ22524/FLJ22622/MGC78448/NOEL1A/RCP/rab11-FIP1
RAB11FIP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
80223
RAB11FIP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKM
RUO
RAB11FIP1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers