missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAB11FIP5 Partial ORF (NP_056285.1, 554 a.a. - 652 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026056-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16175286

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: SGLEKLKTVTSGSIQPVTQAPQAGQMVDTKRLKDSAVLDQSAKYYHLTHDELISLLLQRERELSQRDEHVQELESYIDRLLVRIMETSPTLLQIPPGPP
Specifications

Specifications

NP_056285.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SGLEKLKTVTSGSIQPVTQAPQAGQMVDTKRLKDSAVLDQSAKYYHLTHDELISLLLQRERELSQRDEHVQELESYIDRLLVRIMETSPTLLQIPPGPP
RUO
RAB11FIP5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26056
RAB11FIP5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434H018/GAF1/KIAA0857/RIP11/pp75
RAB11FIP5
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers