missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RAET1G Full-length ORF (AAH98152.1, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH98152.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 353091 |
Molecular Weight (g/mol) | 50.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16120324
|
Abnova™
H00353091-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16110324
|
Abnova™
H00353091-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the RAET1 family, such as RAET1G, are major histocompatibility complex (MHC) class I-related genes located within a 180-kb cluster on chromosome 6q24.2-q25.3. RAET1 proteins contain MHC class I-like alpha-1 and alpha-2 domains. RAET1E (MIM 609243) and RAET1G differ from the other RAET1 proteins (e.g., RAET1I, or ULBP1; MIM 605697) in that they have type I membrane-spanning sequences at their C termini rather than glycosylphosphatidylinositol anchor sequences. (Radosavljevic et al., 2002 [PubMed 11827464]).[supplied by OMIM]
Sequence: MAAAASPAILLRLPLLLLLSSWCRTGLADPHSLCYDITVIPKFRPGPRWRAVQGQVDEKTFLHYDCGSKTVTPVSPLGKKLNVTTAWKAQNPVLREVVDILTEQLLDIQLENYIPKEPLTLQARMSCEQKAEGHGSGSWQLSFDGQIFLLFDSENRMWTTVHPGARKMKEKWENDKDMTMSFHYISMGDCTGWLEDFLMGMDSTLEPSAGGTVSpecifications
AAH98152.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAAASPAILLRLPLLLLLSSWCRTGLADPHSLCYDITVIPKFRPGPRWRAVQGQVDEKTFLHYDCGSKTVTPVSPLGKKLNVTTAWKAQNPVLREVVDILTEQLLDIQLENYIPKEPLTLQARMSCEQKAEGHGSGSWQLSFDGQIFLLFDSENRMWTTVHPGARKMKEKWENDKDMTMSFHYISMGDCTGWLEDFLMGMDSTLEPSAGGTV | |
RUO | |
RAET1G | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
353091 | |
RAET1G (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RAET1G | |
Wheat Germ (in vitro) | |
GST | |
Liquid |