missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAMP2 Partial ORF (NP_005845.1, 58 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010266-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16055225

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the RAMP family of single-transmembrane-domain proteins, called receptor (calcitonin) activity modifying proteins (RAMPs). RAMPs are type I transmembrane proteins with an extracellular N terminus and a cytoplasmic C terminus. RAMPs are required to transport calcitonin-receptor-like receptor (CRLR) to the plasma membrane. CRLR, a receptor with seven transmembrane domains, can function as either a calcitonin-gene-related peptide (CGRP) receptor or an adrenomedullin receptor, depending on which members of the RAMP family are expressed. In the presence of this (RAMP2) protein, CRLR functions as an adrenomedullin receptor. The RAMP2 protein is involved in core glycosylation and transportation of adrenomedullin receptor to the cell surface. [provided by RefSeq]

Sequence: VKNYETAVQFCWNHYKDQMDPIEKDWCDWAMISRPYSTLRDCLEHFAELFDLGFPNPLAERIIFETHQIHFANCSLVQPTFSDPPEDV
Specifications

Specifications

NP_005845.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VKNYETAVQFCWNHYKDQMDPIEKDWCDWAMISRPYSTLRDCLEHFAELFDLGFPNPLAERIIFETHQIHFANCSLVQPTFSDPPEDV
RUO
RAMP2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
10266
RAMP2 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RAMP2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers