Learn More
Abnova™ Human RASD1 Full-length ORF (NP_057168.1, 1 a.a. - 281 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051655-P02.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a Ras-related protein that is stimulated by dexamethasone. The exact function of this gene is unknown, but it may play a role in dexamethasone-induced alterations in cell morphology, growth and cell-extracellular matrix interactions. In addition, studies of a similar rat protein suggest that it functions as as a novel physiologic nitric oxide (NO) effector. The gene product belongs to the Ras superfamily of small GTPases. [provided by RefSeq]
Sequence: MKLAAMIKKMCPSDSELSIPAKNCYRMVILGSSKVGKTAIVSRFLTGRFEDAYTPTIEDFHRKFYSIRGEVYQLDILDTSGNHPFPAMRRLSILTGDVFILVFSLDNRDSFEEVQRLRQQILDTKSCLKNKTKENVDVPLVICGNKGDRDFYREVDQREIEQLVGDDPQRCAYFEISAKKNSSLDQMFRALFAMAKLPSEMSPDLHRKVSVQYCDVLHKKALRNKKLLRAGSGGGGGDPGDAFGIVAPFARRPSVHSDLMYIREKASAGSQAKDKERCVISSpecifications
NP_057168.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGS1/DEXRAS1/MGC:26290 | |
RASD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51655 | |
RASD1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLAAMIKKMCPSDSELSIPAKNCYRMVILGSSKVGKTAIVSRFLTGRFEDAYTPTIEDFHRKFYSIRGEVYQLDILDTSGNHPFPAMRRLSILTGDVFILVFSLDNRDSFEEVQRLRQQILDTKSCLKNKTKENVDVPLVICGNKGDRDFYREVDQREIEQLVGDDPQRCAYFEISAKKNSSLDQMFRALFAMAKLPSEMSPDLHRKVSVQYCDVLHKKALRNKKLLRAGSGGGGGDPGDAFGIVAPFARRPSVHSDLMYIREKASAGSQAKDKERCVIS | |
RUO | |
RASD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |