missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RASGRP3 Partial ORF (NP_733772.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00025780-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16065375

  • 6845.00 SEK / 25µg
Estimated Shipment: 10-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the RAS (see HRAS; MIM 190020) subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation (Rebhun et al., 2000 [PubMed 10934204]).[supplied by OMIM]

Sequence: MGSSGLGKAATLDELLCTCIEMFDDNGELDNSYLPRIVLLMHRWYLSSTELAEKLLCMYRNATGESCNEFRLKICYFMRYWILKFPAEFNLDLGLIRMT
Specifications

Specifications

NP_733772.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSSGLGKAATLDELLCTCIEMFDDNGELDNSYLPRIVLLMHRWYLSSTELAEKLLCMYRNATGESCNEFRLKICYFMRYWILKFPAEFNLDLGLIRMT
RUO
RASGRP3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
25780
RASGRP3 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GRP3/KIAA0846
RASGRP3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers