Learn More
Abnova™ Human RASSF5 Partial ORF (NP_113625, 100 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083593-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the Ras association domain family. It functions as a tumor suppressor, and is inactivated in a variety of cancers. The encoded protein localizes to centrosomes and microtubules, and associates with the GTP-activated forms of Ras, Rap1, and several other Ras-like small GTPases. The protein regulates lymphocyte adhesion and suppresses cell growth in response to activated Rap1 or Ras. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: DVRSIFEQPQDPRVPAERGEGHCFAELVLPGGPGWCDLCGREVLRQALRCTNCKFTCHPECRSLIQLDCSQQEGLSRDRPSPESTLTVTFSQNVCKPVEETQRPPTLSpecifications
NP_113625 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DVRSIFEQPQDPRVPAERGEGHCFAELVLPGGPGWCDLCGREVLRQALRCTNCKFTCHPECRSLIQLDCSQQEGLSRDRPSPESTLTVTFSQNVCKPVEETQRPPTL | |
RUO | |
RASSF5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
83593 | |
RASSF5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC10823/MGC17344/Maxp1/NORE1/NORE1A/NORE1B/RAPL/RASSF3 | |
RASSF5 | |
Recombinant | |
wheat germ expression system |