missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human Rax Partial Orf (Np_038463.2, 104 A.A.-206 A.A.) Recombinant Protein With Gst Tag At N-Terminal.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

4125.00 SEK - 6260.00 SEK

Specifications

Accession Number NP_038463.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 30062
Name RAX (Human) Recombinant Protein (Q01)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16005485
View Documents Promotion Details
Abnova™
H00030062-Q01.10UG
10 ug
4125.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 06-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16015485
View Documents Promotion Details
Abnova™
H00030062-Q01.25UG
25 ug
6260.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 06-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: KEPWEARPSPGLPVGPATGEAKLSEEEQPKKKHRRNRTTFTTYQLHELERAFEKSHYPDVYSREELAGKVNLPEVRVQVWFQNRRAKWRRQEKLEVSSMKLQD

Specifications

Specifications

NP_038463.2
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
RAX (Human) Recombinant Protein (Q01)
12.5% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MCOP3/RX
RAX
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
30062
Glutathione Sepharose 4 Fast Flow
KEPWEARPSPGLPVGPATGEAKLSEEEQPKKKHRRNRTTFTTYQLHELERAFEKSHYPDVYSREELAGKVNLPEVRVQVWFQNRRAKWRRQEKLEVSSMKLQD
RUO
RAX
GST
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only