Learn More
Abnova™ Human RBAK Partial ORF (NP_066986, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057786-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a nuclear protein which interacts with the tumor suppressor retinoblastoma 1. The two interacting proteins are thought to act as a transcriptional repressor for promoters which are activated by the E2F1 transcription factor. This protein contains a Kruppel-associated box (KRAB), which is a transcriptional repressor motif. [provided by RefSeq]
Sequence: TTKPNVIIKLEQGEEPWIMGGEFPCQHSPEAWRVDDLIERIQENEDKHSRQAACINSKTLTEEKENTFSQIYMETSLVPSSIIAHNCVSCGKNLESISQLSpecifications
NP_066986 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TTKPNVIIKLEQGEEPWIMGGEFPCQHSPEAWRVDDLIERIQENEDKHSRQAACINSKTLTEEKENTFSQIYMETSLVPSSIIAHNCVSCGKNLESISQL | |
RUO | |
RBAK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57786 | |
RBAK (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZNF769 | |
RBAK | |
Recombinant | |
wheat germ expression system |