Learn More
Abnova™ Human RBBP6 Full-length ORF (NP_116015.2, 1 a.a. - 118 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005930-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The retinoblastoma tumor suppressor (pRB) protein binds with many other proteins. In various human cancers, pRB suppresses cellular proliferation and is inactivated. Cell cycle-dependent phosphorylation regulates the activity of pRB. This gene encodes a protein which binds to underphosphorylated but not phosphorylated pRB. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSCVHYKFSSKLNYDTVTFDGLHISLCDLKKQIMGREKLKAADCDLQITNAQTKEEYTDDNALIPKNSSVIVRRIPIGGVKSTSKTYVISRTEPAMATTKAVCKNTISHFFYTLLLPLSpecifications
NP_116015.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686P0638/DKFZp761B2423/MY038/P2P-R/PACT/RBQ-1/SNAMA | |
RBBP6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5930 | |
RBBP6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSCVHYKFSSKLNYDTVTFDGLHISLCDLKKQIMGREKLKAADCDLQITNAQTKEEYTDDNALIPKNSSVIVRRIPIGGVKSTSKTYVISRTEPAMATTKAVCKNTISHFFYTLLLPL | |
RUO | |
RBBP6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |