Learn More
Abnova™ Human RBM10 Partial ORF (NP_690595.1, 123 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008241-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3' end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: QKVSMHYSDPKPKINEDWLCNKCGVQNFKRREKCFKCGVPKSEAEQKLPLGTRLDQQTLPLGGRELSQGLLPLPQPYQAQGVLASQALSQGSEPSSENANDTIILRNLSpecifications
NP_690595.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QKVSMHYSDPKPKINEDWLCNKCGVQNFKRREKCFKCGVPKSEAEQKLPLGTRLDQQTLPLGGRELSQGLLPLPQPYQAQGVLASQALSQGSEPSSENANDTIILRNL | |
RUO | |
RBM10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8241 | |
RBM10 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DXS8237E/GPATC9/GPATCH9/KIAA0122/MGC1132/MGC997/ZRANB5 | |
RBM10 | |
Recombinant | |
wheat germ expression system |