missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBM23 Full-length ORF (NP_060577.2, 1 a.a. - 424 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055147-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121183

  • 4515.00 SEK / 10µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the U2AF-like family of RNA binding proteins. This protein interacts with some steroid nuclear receptors, localizes to the promoter of a steroid- responsive gene, and increases transcription of steroid-responsive transcriptional reporters in a hormone-dependent manner. It is also implicated in the steroid receptor-dependent regulation of alternative splicing. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MASDDFDIVIEAMLEAPYKKEEDEQQRKEVKKDYPSNTTSSTSNSGNETSGSSTIGETSNRSRDRDRYRRRNSRSRSPGRQCRHRSRSWDRRHGSESRSRDHRREDRVHYRSPPLATGYRYGHSKSPHFREKSPVREPVDNLSPEERDARTVFCMQLAARIRPRDLEDFFSAVGKVRDVRIISDRNSRRSKGIAYVEFCEIQSVPLAIGLTGQRLLGVPIIVQASQAEKNRLAAMANNLQKGNGGPMRLYVGSLHFNITEDMLRGIFEPFGKIDNIVLMKDSDTGRSKGYGFITFSDSECARRALEQLNGFELAGRPMRVGHVTERLDGGTDITFPDGDQELDLGSAGGRFQLMAKLAEGAGIQLPSTAAAAAAAAAAQAAALQLNGAVPLGALNPAALTALSPALNLASQCFQLSSLFTPQTM
Specifications

Specifications

NP_060577.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAPERbeta/FLJ10482/MGC4458/PP239/RNPC4
RBM23
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55147
RBM23 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASDDFDIVIEAMLEAPYKKEEDEQQRKEVKKDYPSNTTSSTSNSGNETSGSSTIGETSNRSRDRDRYRRRNSRSRSPGRQCRHRSRSWDRRHGSESRSRDHRREDRVHYRSPPLATGYRYGHSKSPHFREKSPVREPVDNLSPEERDARTVFCMQLAARIRPRDLEDFFSAVGKVRDVRIISDRNSRRSKGIAYVEFCEIQSVPLAIGLTGQRLLGVPIIVQASQAEKNRLAAMANNLQKGNGGPMRLYVGSLHFNITEDMLRGIFEPFGKIDNIVLMKDSDTGRSKGYGFITFSDSECARRALEQLNGFELAGRPMRVGHVTERLDGGTDITFPDGDQELDLGSAGGRFQLMAKLAEGAGIQLPSTAAAAAAAAAAQAAALQLNGAVPLGALNPAALTALSPALNLASQCFQLSSLFTPQTM
RUO
RBM23
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers