missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Abnova™ Human RCP9 Partial ORF (NP_055293, 39 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal
Beskrivning
This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: QQNLNTITYETLKYISKTPCRHQSPEIVREFLTALKSHKLTKAEKLQLLNHRPVTAVEIQLMVEESEERLTEEQIEALLHTVTSILPAEPEAEQKKNTNSNVAMDEEDPA
Specifikationer
Specifikationer
| Tillträdesnummer | NP_055293 |
| För användning med (applikation) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulering | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gen-ID (Entrez) | 27297 |
| Molekylvikt (g/mol) | 37.84kDa |
| Namn | RCP9 (Human) Recombinant Protein (Q01) |
| Kvalitetskontrolltestning | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Kvantitet | 25 μg |
| Immunogen | QQNLNTITYETLKYISKTPCRHQSPEIVREFLTALKSHKLTKAEKLQLLNHRPVTAVEIQLMVEESEERLTEEQIEALLHTVTSILPAEPEAEQKKNTNSNVAMDEEDPA |
| Förvaringskrav | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?