missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RDBP Partial ORF (NP_002895.3, 281 a.a. - 379 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_002895.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7936 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128085
|
Abnova™
H00007936-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16118085
|
Abnova™
H00007936-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. [provided by RefSeq]
Sequence: SPFGNIIDLSMDPPRNCAFVTYEKMESADQAVAELNGTQVESVQLKVNIARKQPMLDAATGKSVWGSLAVQNSPKGCHRDKRTQIVYSDDVYKENLVDGSpecifications
NP_002895.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D6S45/NELF-E/RD/RDP | |
RDBP | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7936 | |
RDBP (Human) Recombinant Protein (Q01) | |
SPFGNIIDLSMDPPRNCAFVTYEKMESADQAVAELNGTQVESVQLKVNIARKQPMLDAATGKSVWGSLAVQNSPKGCHRDKRTQIVYSDDVYKENLVDG | |
RUO | |
RDBP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |