Läs mer
STEMCELL Technologies Human Recombinant G-CSF, ACF
Produktkod. 17109661
Encompass_Preferred
Kvantitet:
10 μg
100 μg
1000 μg
missing translation for 'unitSize'
1000µg
100µg
Product Code. 17109661
missing translation for 'mfr': STEMCELL Technologies 78138.1
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Granulocyte colony-stimulating factor, animal component-free
  • Granulocyte colony-stimulating factor (G-CSF) is a member of the CSF family of glycoproteins that regulate hematopoietic cell proliferation, differentiation, and function
  • It is a key cytokine involved in the production of neutrophils and the stimulation of granulocyte colony formation from hematopoietic progenitor cells
  • G-CSF causes a range of effects including a transient reduction of SDF-1 expression, the activation of metalloproteases that cleave VCAM-1, and the release of norepinephrine from the sympathetic nervous system, leading to the release or mobilization of hematopoietic stem cells from the bone marrow into the periphery
  • The G-CSF receptor is expressed on a variety of hematopoietic cells, including myeloid-committed progenitor cells, neutrophils, granulocytes, and monocytes
Specifications
Namn Colony-stimulating factor 3, CSF-3, MGI-1G, Pluripoietin
Kvantitet 100 μg
Celltyp Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neurons
Renhet eller kvalitetsklass ≥95%
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.