Learn More
STEMCELL Technologies Human Recombinant IL-10
Interleukin 10
Brand:  STEMCELL Technologies 78024.1
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17199371
  • 3492.58 SEK / 10µg
Estimated Shipment: 09-06-2023
to see stock.
Description
Description
  • Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines
  • All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds
  • Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3
  • It is produced by dendritic cells, macrophages, and CD4+ T cells, as well as mast cells, NK cells, neutrophils, and B cells, under specific stimulating conditions (Saraiva & O'Garra)
  • IL-10 can inhibit the expression of pro-inflammatory cytokines and promote healing processes, and is important for the function of regulatory T cells
  • IL-10 also enhances B cell proliferation, immunoglobulin secretion, and class II MHC expression, while IL-10 produced by macrophages inhibits activation of neighboring macrophages, thus allowing a level of self-regulation (Ouyang et al.)
Specifications
Specifications
B-TCGF, CSIF, Cytokine synthesis inhibitory factor, Interleukin-10, TGIF
≥95%
10 μg
Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells