Läs mer
STEMCELL Technologies Human Recombinant IL-10
Kvantitet:
10 μg
1000 μg
50 μg
Beskrivning
- Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines
- All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds
- Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3
- It is produced by dendritic cells, macrophages, and CD4+ T cells, as well as mast cells, NK cells, neutrophils, and B cells, under specific stimulating conditions (Saraiva & O'Garra)
- IL-10 can inhibit the expression of pro-inflammatory cytokines and promote healing processes, and is important for the function of regulatory T cells
- IL-10 also enhances B cell proliferation, immunoglobulin secretion, and class II MHC expression, while IL-10 produced by macrophages inhibits activation of neighboring macrophages, thus allowing a level of self-regulation (Ouyang et al.)
Specifikationer
Specifikationer
| Kvantitet | 1000 μg |
| Celltyp | Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells |
| Renhet eller kvalitetsklass | ≥95% |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.