Läs mer
STEMCELL Technologies Human Recombinant IL-15
Kvantitet:
10 μg
100 μg
1000 μg
Beskrivning
- Interleukin 15 (IL-15) is a four-alpha helix bundle cytokine with many similar properties to IL-2, with which it shares components of its receptor
- The IL-15 receptor is a heterotrimeric receptor composed of IL-15Ra, the high-affinity receptor for IL-15, as well as IL-2/15Rb (CD122) and common gamma chain (CD132)
- IL-15 binds to IL-15Rα receptor and can then be presented in trans to IL-2/15Rb and common gamma chain on other cells
- Trans-presentation is thought to be the major mechanism by which IL-15-mediated responses occur in mice, although may not be necessary in humans (Castillo et al.) The cytoplasmic domains of IL-2/15Rb and common gamma chain mediate signaling to activate JAK/STAT and PI3K pathways
- IL-15 supports the survival and proliferation of naive CD4+ and CD8+ T cells, and promotes homeostasis of memory T cells
- IL-15 also promotes the survival and differentiation of NK cells and regulates their cytolytic activity (Ma et al.)
Specifikationer
Specifikationer
| Namn | IL-T, Interleukin-15 |
| Kvantitet | 1000 μg |
| Celltyp | Hematopoietic Stem and Progenitor Cells; Lymphocytes; NK Cells |
| Renhet eller kvalitetsklass | ≥95% |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.