Learn More
STEMCELL Technologies Human Recombinant IL-15, ACF
Interleukin 15, animal component-free
Brand: STEMCELL Technologies 78218.2
Additional Details : Weight : 0.00110kg
Description
- Interleukin 15 (IL-15) is a four-alpha helix bundle cytokine with many similar properties to IL-2
- The IL-15 receptor is a heterotrimeric receptor composed of IL-15Ra (the high-affinity receptor for IL-15), as well as IL-2/15Rb (CD122) and common gamma chain (CD132)
- IL-15 binds to IL-15Rα receptor and can then be presented in trans to IL-2/15Rb and common gamma chain on other cells
- Trans-presentation is thought to be the major mechanism by which IL-15-mediated responses occur in mice, although may not be necessary in humans (Castillo et al.) The cytoplasmic domains of IL-2/15Rb and common gamma chain mediate signaling to activate JAK/STAT and PI3K pathways
- IL-15 supports the survival and proliferation of naïve CD4+ and CD8+ T cells, and promotes homeostasis of memory T cells
- IL-15 also promotes the survival and differentiation of NK cells and regulates their cytolytic activity (Ma et al.) This product is animal component-free
Specifications
IL-T, Interleukin-15 | |
≥95% | |
500 μg | |
Hematopoietic Stem and Progenitor Cells; Lymphocytes; NK Cells |