missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RFC4 Full-length ORF (NP_002907.1, 1 a.a. - 363 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005984-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122238

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 37 kD subunit. This subunit forms a core complex with the 36 and 40 kDa subunits. The core complex possesses DNA-dependent ATPase activity, which was found to be stimulated by PCNA in an in vitro system. Alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]

Sequence: MQAFLKGTSISTKPPLTKDRGVAASAGSSGENKKAKPVPWVEKYRPKCVDEVAFQEEVVAVLKKSLEGADLPNLLFYGPPGTGKTSTILAAARELFGPELFRLRVLELNASDERGIQVVREKVKNFAQLTVSGSRSDGKPCPPFKIVILDEADSMTSAAQAALRRTMEKESKTTRFCLICNYVSRIIEPLTSRCSKFRFKPLSDKIQQQRLLDIAKKENVKISDEGIAYLVKVSEGDLRKAITFLQSATRLTGGKEITEKVITDIAGVIPAEKIDGVFAACQSGSFDKLEAVVKDLIDEGHAATQLVNQLHDVVVENNLSDKQKSIITEKLAEVDKCLADGADEHLQLISLCATVMQQLSQNC
Specifications

Specifications

NP_002907.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A1/MGC27291/RFC37
RFC4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5984
RFC4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQAFLKGTSISTKPPLTKDRGVAASAGSSGENKKAKPVPWVEKYRPKCVDEVAFQEEVVAVLKKSLEGADLPNLLFYGPPGTGKTSTILAAARELFGPELFRLRVLELNASDERGIQVVREKVKNFAQLTVSGSRSDGKPCPPFKIVILDEADSMTSAAQAALRRTMEKESKTTRFCLICNYVSRIIEPLTSRCSKFRFKPLSDKIQQQRLLDIAKKENVKISDEGIAYLVKVSEGDLRKAITFLQSATRLTGGKEITEKVITDIAGVIPAEKIDGVFAACQSGSFDKLEAVVKDLIDEGHAATQLVNQLHDVVVENNLSDKQKSIITEKLAEVDKCLADGADEHLQLISLCATVMQQLSQNC
RUO
RFC4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers