missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RFP2 Full-length ORF (NP_001007279.1, 1 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010206-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16182106

  • 5030.41 SEK / 25µg

Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This gene is located on chromosome 13 within the minimal deletion region for B-cell chronic lymphocytic leukemia. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]

Sequence: MDVMELLEEDLTCPICCSLFDDPRVLPCSHNFCKKCLEGILEGSVRNSLWRPAPFKCPTCRKETSATGINSLQVNYSLKGIVEKYNKIKISPKMPVCKGHLGQPLNIFCLTDMQLICGICATRGEHTKHVFCSIEDAYAQERDAFESLFQSFETWRRGDALSRLDTLETSKRKSLQLLTKDSDKVKEFFEKLQHTLDQKKNEILSDFETMKLAVMQAYDPEINKLNTILQEQRMAFNIAEAFKDVSEPIVFLQQMQEFREKIKVIKETPLPPSNLPASPLMKNFDTSQWEDIKLVDVDKLSLPQDTGTFISKIPWSFYKLFLLILLLGLVIVFGPTMFLEWSLFDDLATWKGCLSNFSSYLTKTADFIEQSVFYWEQVTDGFFIFNERFKNFTLVVLNNVAEFVCKYKLL
Specifications

Specifications

NP_001007279.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAR/DLEU5/LEU5/RFP2/RNF77
TRIM13
Yes
wheat germ expression system
Liquid
10206
RFP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDVMELLEEDLTCPICCSLFDDPRVLPCSHNFCKKCLEGILEGSVRNSLWRPAPFKCPTCRKETSATGINSLQVNYSLKGIVEKYNKIKISPKMPVCKGHLGQPLNIFCLTDMQLICGICATRGEHTKHVFCSIEDAYAQERDAFESLFQSFETWRRGDALSRLDTLETSKRKSLQLLTKDSDKVKEFFEKLQHTLDQKKNEILSDFETMKLAVMQAYDPEINKLNTILQEQRMAFNIAEAFKDVSEPIVFLQQMQEFREKIKVIKETPLPPSNLPASPLMKNFDTSQWEDIKLVDVDKLSLPQDTGTFISKIPWSFYKLFLLILLLGLVIVFGPTMFLEWSLFDDLATWKGCLSNFSSYLTKTADFIEQSVFYWEQVTDGFFIFNERFKNFTLVVLNNVAEFVCKYKLL
RUO
TRIM13
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates