missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RFXAP Full-length ORF (AAH26088.1, 1 a.a. - 272 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005994-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180192

  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated ankyrin-containing protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group D. Transcript variants utilizing different polyA signals have been found for this gene. [provided by RefSeq]

Sequence: MEAQSVAEGAGPGAASGVPHPAALAPAAAPTLAPASVAAAASQFTLLVMQPCAGQDEAAAPGGSVGAGKPVRYLCEGAGDGEEEAGEDEADLLDTSDPPGGGESAASLEDLEDEETHSGGEGSSGGARRRGSGGGSMSKTCTYEGCSETTSQVAKQRKPWMCKKHRNKMYKDKYKKKKSDQALNCGGTASTGSAGNVKLEESADNILSIVKQRTGSFGDRPARPTLLEQVLNQKRLSLLRSPEVVQFLQKQQQLLNQQVLEQRQQQFPGTSM
Specifications

Specifications

AAH26088.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RFXAP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
5994
RFXAP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEAQSVAEGAGPGAASGVPHPAALAPAAAPTLAPASVAAAASQFTLLVMQPCAGQDEAAAPGGSVGAGKPVRYLCEGAGDGEEEAGEDEADLLDTSDPPGGGESAASLEDLEDEETHSGGEGSSGGARRRGSGGGSMSKTCTYEGCSETTSQVAKQRKPWMCKKHRNKMYKDKYKKKKSDQALNCGGTASTGSAGNVKLEESADNILSIVKQRTGSFGDRPARPTLLEQVLNQKRLSLLRSPEVVQFLQKQQQLLNQQVLEQRQQQFPGTSM
RUO
RFXAP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers