Learn More
Abnova™ Human RGS12 Full-length ORF (ENSP00000304459, 1 a.a. - 555 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006002-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the 'regulator of G protein signaling' (RGS) gene family. The encoded protein may function as a guanosine triphosphatase (GTPase)-activating protein as well as a transcriptional repressor. This protein may play a role in tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their biological nature has not been determined. [provided by RefSeq]
Sequence: MKKHDFAENLPSSLLIIILVIFTLSGSKFGRGTGLTQPSQRTSARRSFGRSKRFSITRSLDDLESATVSDGELTGADLKDCVSNNSLSSNASLPSVQSCRRLRERRVASWAVSFERLLQDPVGVRYFSDFLRKEFSEENILFWQACEYFNHVPAHDKKELSYRAREIFSKFLCSKATTPVNIDSQAQLADDVLRAPHPDMFKEQQLQIFNLMKFDSYTRFLKSPLYQECILAEVEGRALPDSQQVPSSPASKHSLGSDHSSVSTPKKLSGKSKSGRSLNEELGDEDSEKKRKGAFFSWSRTRSTGRSQKKREHGDHADDALHANGGLCRRESQGSVSSAGSLDLSEACRTLAPEKDKATKHCCIHLPDGTSCVVAVKAGFSIKDILSGLCERHGINGAAADLFLVGGDKPLVLHQDSSILESRDLRLEKRTLFRLDLVPINRSVGLKAKPTKPVTEVLRPVVARYGLDLSGLLVRLSGEKEPLDLGAPISSLDGQRVVLEEKDPSRGKASADKQKGVPVKQNTAVNSSSRNHSATSFLSLFPKLRATEQMTNVGCSpecifications
ENSP00000304459 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761K1617/DKFZp761K1817 | |
RGS12 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6002 | |
RGS12 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKKHDFAENLPSSLLIIILVIFTLSGSKFGRGTGLTQPSQRTSARRSFGRSKRFSITRSLDDLESATVSDGELTGADLKDCVSNNSLSSNASLPSVQSCRRLRERRVASWAVSFERLLQDPVGVRYFSDFLRKEFSEENILFWQACEYFNHVPAHDKKELSYRAREIFSKFLCSKATTPVNIDSQAQLADDVLRAPHPDMFKEQQLQIFNLMKFDSYTRFLKSPLYQECILAEVEGRALPDSQQVPSSPASKHSLGSDHSSVSTPKKLSGKSKSGRSLNEELGDEDSEKKRKGAFFSWSRTRSTGRSQKKREHGDHADDALHANGGLCRRESQGSVSSAGSLDLSEACRTLAPEKDKATKHCCIHLPDGTSCVVAVKAGFSIKDILSGLCERHGINGAAADLFLVGGDKPLVLHQDSSILESRDLRLEKRTLFRLDLVPINRSVGLKAKPTKPVTEVLRPVVARYGLDLSGLLVRLSGEKEPLDLGAPISSLDGQRVVLEEKDPSRGKASADKQKGVPVKQNTAVNSSSRNHSATSFLSLFPKLRATEQMTNVGC | |
RUO | |
RGS12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |