missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RHCG Partial ORF (NP_057405, 418 a.a. - 479 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051458-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117016

  • 3860.70 SEK / 10µg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LPFWGQPSDENCFEDAVYWEMPEGNSTVYIPEDPTFKPSGPSVPSVPMVSPLPMASSVPLVP
Specifications

Specifications

NP_057405
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.56kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LPFWGQPSDENCFEDAVYWEMPEGNSTVYIPEDPTFKPSGPSVPSVPMVSPLPMASSVPLVP
RUO
RHCG
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
51458
RHCG (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C15orf6/PDRC2/RHGK/SLC42A3
RHCG
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers