missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RIC8A Partial ORF (NP_068751.4, 462 a.a. - 534 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00060626-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179446

  • 5030.41 SEK / 25µg

Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: VTGRVEEKPPNPMEGMTEEQKEHEAMKLVTMFDKLSRNRVIQPMGMSPRGHLTSLQDAMCETMEQQLSSDPDS
Specifications

Specifications

NP_068751.4
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VTGRVEEKPPNPMEGMTEEQKEHEAMKLVTMFDKLSRNRVIQPMGMSPRGHLTSLQDAMCETMEQQLSSDPDS
RUO
RIC8A
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
60626
RIC8A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC104517/MGC131931/MGC148073/MGC148074/RIC8/synembryn
RIC8A
Yes
Videos
SDS
Documents
Certificates