missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RIPK2 Partial ORF (AAH04553, 431 a.a. - 540 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008767-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16119205

  • 5854.80 SEK / 25µg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases. The encoded protein contains a C-terminal caspase activation and recruitment domain (CARD), and is a component of signaling complexes in both the innate and adaptive immune pathways. It is a potent activator of NF-kappaB and inducer of apoptosis in response to various stimuli. [provided by RefSeq]

Sequence: LQPGIAQQWIQSKREDIVNQMTEACLNQSLDALLSRDLIMKEDYELVSTKPTRTSKVRQLLDTTDIQGEEFAKVIVQKLKDNKQMGLQPYPEILVVSRSPSLNLLQNKSM
Specifications

Specifications

AAH04553
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LQPGIAQQWIQSKREDIVNQMTEACLNQSLDALLSRDLIMKEDYELVSTKPTRTSKVRQLLDTTDIQGEEFAKVIVQKLKDNKQMGLQPYPEILVVSRSPSLNLLQNKSM
RUO
RIPK2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
8767
RIPK2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CARD3/CARDIAK/CCK/GIG30/RICK/RIP2
RIPK2
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers