missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF111 Partial ORF (NP_060080, 1 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_060080 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54778 |
Molecular Weight (g/mol) | 37.62kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16167736
|
Abnova™
H00054778-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16177736
|
Abnova™
H00054778-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene contains a RING finger domain, a motif known to be involved in protein-protein and protein-DNA interactions. The mouse counterpart of this gene (Rnf111/arkadia) has been shown to genetically interact with the transforming growth factor (TGF) beta-like factor Nodal, and act as a modulator of the nodal signaling cascade, which is essential for the induction of mesoderm during embryonic development. [provided by RefSeq]
Sequence: MSQWTPEYNELYTLKVDMKSEIPSDAPKTQESLKGILLHPEPIGAAKSFPAGVEMINSKVGNEFSHLCDDSQKQEKEMNGNQQEQEKSLVVRKKRKSQQAGPSYVQNCSpecifications
NP_060080 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARK/DKFZp313E0731/DKFZp686H1966/DKFZp761D081/FLJ38008 | |
RNF111 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
54778 | |
RNF111 (Human) Recombinant Protein (Q01) | |
MSQWTPEYNELYTLKVDMKSEIPSDAPKTQESLKGILLHPEPIGAAKSFPAGVEMINSKVGNEFSHLCDDSQKQEKEMNGNQQEQEKSLVVRKKRKSQQAGPSYVQNC | |
RUO | |
RNF111 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |