missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF167 Full-length ORF (NP_056343.1, 1 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_056343.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26001 |
Molecular Weight (g/mol) | 64.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16147992
|
Abnova™
H00026001-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137992
|
Abnova™
H00026001-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RNF167 is an E3 ubiquitin ligase that interacts with TSSC5 (SLC22A18; MIM 602631) and, together with UBCH6 (UBE2E1; MIM 602916), facilitates TSSC5 polyubiquitylation (Yamada and Gorbsky, 2006 [PubMed 16314844]).[supplied by OMIM]
Sequence: MHPAAFPLPVVVAAVLWGAAPTRGLIRATSDHNASMDFADLPALFGATLSQEGLQGFLVEAHPDNACSPIAPPPPAPVNGSVFIALLRRFDCNFDLKVLNAQKAGYGAAVVHNVNSNELLNMVWNSEEIQQQIWIPSVFIGERSSEYLRALFVYEKGARVLLVPDNTFPLGYYLIPFTGIVGLLVLAMGAVMIARCIQHRKRLQRNRLTKEQLKQIPTHDYQKGDQYDVCAICLDEYEDGDKLRVLPCAHAYHSRCVDPWLTQTRKTCPICKQPVHRGPGDEDQEEETQGQEEGDEGEPRDHPASERTPLLGSSPTLPTSFGSLAPAPLVFPGPSTDPPLSPPSSPVILVSpecifications
NP_056343.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MHPAAFPLPVVVAAVLWGAAPTRGLIRATSDHNASMDFADLPALFGATLSQEGLQGFLVEAHPDNACSPIAPPPPAPVNGSVFIALLRRFDCNFDLKVLNAQKAGYGAAVVHNVNSNELLNMVWNSEEIQQQIWIPSVFIGERSSEYLRALFVYEKGARVLLVPDNTFPLGYYLIPFTGIVGLLVLAMGAVMIARCIQHRKRLQRNRLTKEQLKQIPTHDYQKGDQYDVCAICLDEYEDGDKLRVLPCAHAYHSRCVDPWLTQTRKTCPICKQPVHRGPGDEDQEEETQGQEEGDEGEPRDHPASERTPLLGSSPTLPTSFGSLAPAPLVFPGPSTDPPLSPPSSPVILV | |
RUO | |
RNF167 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26001 | |
RNF167 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
5730408C10Rik/DKFZp566H073/LP2254/RING105 | |
RNF167 | |
Recombinant | |
wheat germ expression system |