Learn More
Abnova™ Human RNPS1 Full-length ORF (NP_006702.1, 1 a.a. - 305 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006702.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10921 |
Molecular Weight (g/mol) | 60.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135862
|
Abnova™
H00010921-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125862
|
Abnova™
H00010921-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. This protein contains many serine residues. Two splice variants have been found for this gene; both variants encode the same protein. [provided by RefSeq]
Sequence: MDLSGVKKKSLLGVKENNKKSSTRAPSPTKRKDRSDEKSKDRSKDKGATKESSEKDRGRDKTRKRRSASSGSSSTRSRSSSTSSSGSSTSTGSSSGSSSSSASSRSGSSSTSRSSSSSSSSGSPSPSRRRHDNRRRSRSKSKPPKRDEKERKRRSPSPKPTKVHIGRLTRNVTKDHIMEIFSTYGKIKMIDMPVERMHPHLSKGYAYVEFENPDEAEKALKHMDGGQIDGQEITATAVLAPWPRPPPRRFSPPRRMLPPPPMWRRSPPRMRRRSRSPRRRSPVRRRSRSPGRRRHRSRSSSNSSRSpecifications
NP_006702.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDLSGVKKKSLLGVKENNKKSSTRAPSPTKRKDRSDEKSKDRSKDKGATKESSEKDRGRDKTRKRRSASSGSSSTRSRSSSTSSSGSSTSTGSSSGSSSSSASSRSGSSSTSRSSSSSSSSGSPSPSRRRHDNRRRSRSKSKPPKRDEKERKRRSPSPKPTKVHIGRLTRNVTKDHIMEIFSTYGKIKMIDMPVERMHPHLSKGYAYVEFENPDEAEKALKHMDGGQIDGQEITATAVLAPWPRPPPRRFSPPRRMLPPPPMWRRSPPRMRRRSRSPRRRSPVRRRSRSPGRRRHRSRSSSNSSR | |
RUO | |
RNPS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10921 | |
RNPS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
E5.1/MGC117332 | |
RNPS1 | |
Recombinant | |
wheat germ expression system |