Learn More
Abnova™ Human RPA4 Partial ORF (NP_037479.1, 84 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029935-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
Replication protein A (RPA) is an essential factor for DNA double-strand break repair and cell cycle checkpoint activation. This gene encodes the 32-kDa subunit of the RPA, which associates with the 70- and 13-kDa subunits to form a trimeric RPA complex. [provided by RefSeq]
Sequence: EKASNHICYKIDDMTAKPIEARQWFGREKVKQVTPLSVGVYVKVFGILKCPTGTKSLEVLKIHVLEDMNEFTVHILETVNAHMMLDKARRDTTVESVSpecifications
NP_037479.1 | |
Liquid | |
29935 | |
RPA4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSU24186/MGC120333/MGC120334 | |
RPA4 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EKASNHICYKIDDMTAKPIEARQWFGREKVKQVTPLSVGVYVKVFGILKCPTGTKSLEVLKIHVLEDMNEFTVHILETVNAHMMLDKARRDTTVESV | |
RUO | |
RPA4 | |
Wheat Germ (in vitro) | |
GST |