missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RPL3L Partial ORF (NP_005052, 280 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006123-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16144525

  • 6845.00 SEK / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that shares sequence similarity with ribosomal protein L3. The protein belongs to the L3P family of ribosomal proteins. Unlike the ubiquitous expression of ribosomal protein genes, this gene has a tissue-specific pattern of expression, with the highest levels of expression in skeletal muscle and heart. It is not currently known whether the encoded protein is a functional ribosomal protein or whether it has evolved a function that is independent of the ribosome. [provided by RefSeq]

Sequence: LNKKIFRIGRGPHMEDGKLVKNNASTSYDVTAKSITPLGGFPHYGEVNNDFVMLKGCIAGTKKRVITLRKSLLVHHSRQAV
Specifications

Specifications

NP_005052
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LNKKIFRIGRGPHMEDGKLVKNNASTSYDVTAKSITPLGGFPHYGEVNNDFVMLKGCIAGTKKRVITLRKSLLVHHSRQAV
RUO
RPL3L
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
6123
RPL3L (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPL3L
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers