missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RPL41 Full-length ORF (-, 1 a.a. - 25 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006171-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180312

  • 5030.41 SEK / 25µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which shares sequence similarity with the yeast ribosomal protein YL41, belongs to the L41E family of ribosomal proteins. It is located in the cytoplasm. The protein can interact with the beta subunit of protein kinase CKII and can stimulate the phosphorylation of DNA topoisomerase II-alpha by CKII. Two alternative splice variants have been identified, both encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]

Sequence: MRAKWRKKRMRRLKRKRRKMRQRSK
Specifications

Specifications

wheat germ expression system
Liquid
6171
RPL41 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRAKWRKKRMRRLKRKRRKMRQRSK
RUO
RPL41
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
28.49kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPL41
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates