missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RPS6KA2 Partial ORF (AAH02363, 631 a.a. - 733 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH02363 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6196 |
Molecular Weight (g/mol) | 37.07kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184675
|
Abnova™
H00006196-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174675
|
Abnova™
H00006196-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: YALSGGNWDSISDAAKDVVSKMLHVDPHQRLTAMQVLKHPWVVNREYLSPNQLSRQDVHLVKGAMAATYFALNRTPQAPRLEPVLSSNLAQRRGMKRLTSTRLSpecifications
AAH02363 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HU-2/MAPKAPK1C/RSK/RSK3/S6K-alpha/S6K-alpha2/p90-RSK3/pp90RSK3 | |
RPS6KA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6196 | |
RPS6KA2 (Human) Recombinant Protein (Q01) | |
YALSGGNWDSISDAAKDVVSKMLHVDPHQRLTAMQVLKHPWVVNREYLSPNQLSRQDVHLVKGAMAATYFALNRTPQAPRLEPVLSSNLAQRRGMKRLTSTRL | |
RUO | |
RPS6KA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |