Learn More
Abnova™ Human RPS8 Full-length ORF (NP_001003.1, 1 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001003.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6202 |
Molecular Weight (g/mol) | 50.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16190352
|
Abnova™
H00006202-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180352
|
Abnova™
H00006202-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal tumors and colon polyps compared to matched normal colonic mucosa has been observed. This gene is co-transcribed with the small nucleolar RNA genes U38A, U38B, U39, and U40, which are located in its fourth, fifth, first, and second introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]
Sequence: MGISRDNWHKRRKTGGKRKPYHKKRKYELGRPAANTKIGPRRIHTVRVRGGNKKYRALRLDVGNFSWGSECCTRKTRIIDVVYNASNNELVRTKTLVKNCIVLIDSTPYRQWYESHYALPLGRKKGAKLTPEEEEILNKKRSKKIQKKYDERKKNAKISSLLEEQFQQGKLLACIASRPGQCGRADGYVLEGKELEFYLRKIKARKGKSpecifications
NP_001003.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGISRDNWHKRRKTGGKRKPYHKKRKYELGRPAANTKIGPRRIHTVRVRGGNKKYRALRLDVGNFSWGSECCTRKTRIIDVVYNASNNELVRTKTLVKNCIVLIDSTPYRQWYESHYALPLGRKKGAKLTPEEEEILNKKRSKKIQKKYDERKKNAKISSLLEEQFQQGKLLACIASRPGQCGRADGYVLEGKELEFYLRKIKARKGK | |
RUO | |
RPS8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6202 | |
RPS8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPS8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |