missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RSHL1 Partial ORF (NP_110412, 21 a.a. - 113 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_110412 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 81492 |
Molecular Weight (g/mol) | 35.97kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180387
|
Abnova™
H00081492-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16170387
|
Abnova™
H00081492-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is similar to a sea urchin radial spoke head protein. Radial spoke protein complexes form part of the axoneme of eukaryotic flagella and are located between the axoneme's outer ring of doublet microtubules and central pair of microtubules. In Chlamydomonas, radial spoke proteins are thought to regulate the activity of dynein and the symmetry of flagellar bending patterns. This gene maps to a region of chromosome 19 that is linked to primary ciliary dyskinesia-2 (CILD2). [provided by RefSeq]
Sequence: SQASQRRHSRDQAQALAADPEERQQIPPDAQRNAPGWSQRGSLSQQENLLMPQVFQAEEARLGGMEYPSVNTGFPSEFQPQPYSDESRMQVAESpecifications
NP_110412 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.97kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RSP4/RSP6 | |
RSHL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
81492 | |
RSHL1 (Human) Recombinant Protein (Q01) | |
SQASQRRHSRDQAQALAADPEERQQIPPDAQRNAPGWSQRGSLSQQENLLMPQVFQAEEARLGGMEYPSVNTGFPSEFQPQPYSDESRMQVAE | |
RUO | |
RSHL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |