missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RTCD1 Full-length ORF (NP_003720.1, 1 a.a. - 366 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008634-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132702

  • 5030.41 SEK / 25µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RNA 3-prime-terminal phosphate cyclase (RPC; EC 6.5.1.4) catalyzes the ATP-dependent conversion of a 3-prime phosphate to a 2-prime,3-prime-cyclic phosphodiester at the end of RNA (Genschik et al., 1997 [PubMed 9184239]).[supplied by OMIM]

Sequence: MAGPRVEVDGSIMEGGGQILRVSTALSCLLGLPLRVQKIRAGRSTPGLRPQHLSGLEMIRDLCDGQLEGAEIGSTEITFTPEKIKGGIHTADTKTAGSVCLLMQVSMPCVLFAASPSELHLKGGTNAEMAPQIDYTVMVFKPIVEKFGFIFNCDIKTRGYYPKGGGEVIVRMSPVKQLNPINLTERGCVTKIYGRAFVAGVLPFKVAKDMAAAAVRCIRKEIRDLYVNIQPVQEPKDQAFGNGNGIIIIAETSTGCLFAGSSLGKRGVNADKVGIEAAEMLLANLRHGGTVDEYLQDQLIVFMALANGVSRIKTGPVTLHTQTAIHFAEQIAKAKFIVKKSEDEEDAAKDTYIIECQGIGMTNPNL
Specifications

Specifications

NP_003720.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPC
RTCD1
Yes
wheat germ expression system
Liquid
8634
RTCD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGPRVEVDGSIMEGGGQILRVSTALSCLLGLPLRVQKIRAGRSTPGLRPQHLSGLEMIRDLCDGQLEGAEIGSTEITFTPEKIKGGIHTADTKTAGSVCLLMQVSMPCVLFAASPSELHLKGGTNAEMAPQIDYTVMVFKPIVEKFGFIFNCDIKTRGYYPKGGGEVIVRMSPVKQLNPINLTERGCVTKIYGRAFVAGVLPFKVAKDMAAAAVRCIRKEIRDLYVNIQPVQEPKDQAFGNGNGIIIIAETSTGCLFAGSSLGKRGVNADKVGIEAAEMLLANLRHGGTVDEYLQDQLIVFMALANGVSRIKTGPVTLHTQTAIHFAEQIAKAKFIVKKSEDEEDAAKDTYIIECQGIGMTNPNL
RUO
RTCD1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates