Learn More
Abnova™ Human RXRA Partial ORF (NP_002948.1, 27 a.a. - 94 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006256-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
Retinoid X receptors (RXRs) and retinoic acid receptors (RARs), are nuclear receptors that mediate the biological effects of retinoids by their involvement in retinoic acid-mediated gene activation. These receptors exert their action by binding, as homodimers or heterodimers, to specific sequences in the promoters of target genes and regulating their transcription. The protein encoded by this gene is a member of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. [provided by RefSeq]
Sequence: SMAAPSLHPSLGPGIGSPGQLHSPISTLSSPINGMGPPFSVISSPMGPHSMSVPTTPTLGFSTGSPQLSpecifications
NP_002948.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
SMAAPSLHPSLGPGIGSPGQLHSPISTLSSPINGMGPPFSVISSPMGPHSMSVPTTPTLGFSTGSPQL | |
RUO | |
RXRA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6256 | |
RXRA (Human) Recombinant Protein | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ00280/FLJ00318/FLJ16020/FLJ16733/MGC102720/NR2B1 | |
RXRA | |
Recombinant | |
wheat germ expression system |