missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RXRA Partial ORF (NP_002948.1, 27 a.a. - 94 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006256-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: SMAAPSLHPSLGPGIGSPGQLHSPISTLSSPINGMGPPFSVISSPMGPHSMSVPTTPTLGFSTGSPQLSpecifications
NP_002948.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
SMAAPSLHPSLGPGIGSPGQLHSPISTLSSPINGMGPPFSVISSPMGPHSMSVPTTPTLGFSTGSPQL | |
RUO | |
RXRA | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
6256 | |
RXRA (Human) Recombinant Protein | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ00280|FLJ00318|FLJ16020|FLJ16733|MGC102720|NR2B1 | |
RXRA | |
Yes |