missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human S100A6 Partial ORF (NP_055439, 18 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006277-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134855

  • 3343.96 SEK / 10µg

Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma. [provided by RefSeq]

Sequence: KYSGREGDKHTLSKKELKELIQKELTIGSKLQDAEIARLMEDLDRNKDQEVNFQEYVTFLGALALIYSEALKG
Specifications

Specifications

NP_055439
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KYSGREGDKHTLSKKELKELIQKELTIGSKLQDAEIARLMEDLDRNKDQEVNFQEYVTFLGALALIYSEALKG
RUO
S100A6
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
6277
S100A6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
2A9/5B10/CABP/CACY/PRA
S100A6
Yes
Videos
SDS
Documents
Certificates