missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human S100A7 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4475
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human samples. It is validated for Immunohistochemistry.
Specifikationer
| S100A7/Psoriasin | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P31151 | |
| S100A7 | |
| E. coli-derived recombinant human S100A7 Met1-Gln101 Accession # P31151 | |
| 100 μg | |
| Primary | |
| Detects human S100A7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) S100A1, A2, A4, A6, A8, A9, A10, A11, A14, A16, rhS100B, rhS100P, recombinant mouse S100A1, A4, A6, A8, A9, A10, A11, A13, or A16 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 577513 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6278 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering