missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ s90K/Mac-2BP (Soluble) Human ELISA Kit

Produktkod. 15561787
Klicka för att se tillgängliga alternativ
Kvantitet:
96 tester
Förpackningsstorlek:
96 tester
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15561787

Brand: Invitrogen™ BMS234

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Innehåller

  • Aluminum pouch(es) with a Microwell Plate coated with Monoclonal Antibody (murine) to human 90K/Mac-2 BP
  • HRP-Conjugate anti-90K/Mac-2 BP monoclonal (murine) antibody
  • 90K/Mac-2 BP Standard, 200ng/ml; 100ng/ml;50ng/ml;25ng/ml; 12.5ng/ml
  • Wash Buffer Concentrate 10x (phosphate-buffered saline with Tween 20)
  • Sample diluent 10x (buffered protein matrix)
  • Substrate Solution (tetramethyl-benzidine and hydrogen peroxide, stabilized)
  • Stop Solution (0.3M Sulfuric Acid)
  • Adhesive Plate Covers
  • Reagent Labels

Sandwich ELISA Kit

The Human Soluble 90K/Mac-2BP (Hu s90K/Mac-2BP) ELISA quantitates Hu s90K/Mac-2BP in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu s90K/Mac-2BP. Principle of the method The Human s90K/Mac-2BP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer Q08380
Analysintervall 12.5-200 ng/mL
Analyskänslighet 0.92 ng/mL
Konjugera HRP
Produkttyp ELISA Kit
Provtyp Plasma, Serum, Supernatant
Målarter Human
För användning med (applikation) ELISA
För användning med (utrustning) Colorimetric Microplate Reader
Gen-ID (Entrez) 3959
Gensymbol 90K,BTBD17B,CyCAP,M2BP,MAC-2-BP,TANGO10B,gp90
Interassay CV 11.9%
Intraassay CV 5.5%
Kit Innehåll Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
Etikett eller färgämne HRP
Kvantitet 96 tester
Regulatorisk status RUO
Provvolym Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL
Förvaringskrav 2°C to 8°C
Synonym Galectin-3-binding protein, Mac-2-binding protein
Mål Human
Testtid 1 hr. 20 min.
Total analystid 1 hr. 45 min.
Visa mer Visa mindre

For Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.