missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SBF2 Partial ORF (NP_112224, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_112224 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 81846 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130437
|
Abnova™
H00081846-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140437
|
Abnova™
H00081846-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a pseudophosphatase and member of the myotubularin-related protein family. This gene maps within the CMT4B2 candidate region of chromosome 11p15 and mutations in this gene have been associated with Charcot-Marie-Tooth Disease, type 4B2. [provided by RefSeq]
Sequence: MARLADYFIVVGYDHEKPGSGEGLGKIIQRFPQKDWDDTPFPQGIELFCQPGGWQLSRERKQPTFFVVVLTDIDSDRHYCSCLTFYEAEINLQGTKKEEISpecifications
NP_112224 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMT4B2/DKFZp779B2327/FLJ22918/FLJ41627/KIAA1766/MTMR13 | |
SBF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
81846 | |
SBF2 (Human) Recombinant Protein (Q01) | |
MARLADYFIVVGYDHEKPGSGEGLGKIIQRFPQKDWDDTPFPQGIELFCQPGGWQLSRERKQPTFFVVVLTDIDSDRHYCSCLTFYEAEINLQGTKKEEI | |
RUO | |
SBF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |