missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SCTR Full-length ORF (AAH35757.2, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH35757.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6344 |
Molecular Weight (g/mol) | 76.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16170552
|
Abnova™
H00006344-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180552
|
Abnova™
H00006344-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism. [provided by RefSeq]
Sequence: MRPHLSPPLQQLLLPVLLACAAHSTGALPRLCDVLQVLWEEQDQCLQELSREQTGDLGTEQPVPGCEGMWDNISCWPSSVPGRMVEVECPRFLRMLTSRNGSLFRNCTQDGWSETFPRPNLACGVNVNDSSNEKRHSYLLKLKVMYTVGYSSSLVMLLVALGILCAFRRLHCTRNYIHMHLFVSFILRALSNFIKDAVLFSSDDVTYCDAHRAGCKLIMVLFQYCIMANYSWLLVEGLYLHTLLAISFFSERKYLQGFVAFGWGSPAIFVALWAIARHFLEDVGCWDINANASIWWIIRGPVILSILINFILFINILRILMRKLRTQETRGNEVSHYKRLARSTLLLIPLFGIHYIVFAFSPEDAMEIQLFFELALGSFQGLVVAVLYCFLNGEVQLEVQKKWQQWHLREFPLHPVASFSNSTKASHLEQSQGTCRTSIISpecifications
AAH35757.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MRPHLSPPLQQLLLPVLLACAAHSTGALPRLCDVLQVLWEEQDQCLQELSREQTGDLGTEQPVPGCEGMWDNISCWPSSVPGRMVEVECPRFLRMLTSRNGSLFRNCTQDGWSETFPRPNLACGVNVNDSSNEKRHSYLLKLKVMYTVGYSSSLVMLLVALGILCAFRRLHCTRNYIHMHLFVSFILRALSNFIKDAVLFSSDDVTYCDAHRAGCKLIMVLFQYCIMANYSWLLVEGLYLHTLLAISFFSERKYLQGFVAFGWGSPAIFVALWAIARHFLEDVGCWDINANASIWWIIRGPVILSILINFILFINILRILMRKLRTQETRGNEVSHYKRLARSTLLLIPLFGIHYIVFAFSPEDAMEIQLFFELALGSFQGLVVAVLYCFLNGEVQLEVQKKWQQWHLREFPLHPVASFSNSTKASHLEQSQGTCRTSII | |
RUO | |
SCTR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6344 | |
SCTR (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SR | |
SCTR | |
Recombinant | |
wheat germ expression system |