missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SDCCAG10 Full-length ORF (AAH12117.1, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010283-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134812

  • 4055.00 SEK / 10µg
Estimated Shipment: 03-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MANAGSHDNGSQFFFTLGRADELNNKHTIFGKVTGDTVYNMLRLSEVDIDDDERPHNPHKIKSCEVLFNPFDDIIPREIKRLKKEKPEEEVKKLKPKGTKNFSLLSFGEEAEEEEEEVNRVSQSMKGKSKSSHDLLKDDPHLSSVPVVESEKGDAPDLVDDGEDESAEHDEYIDGDEKNLMRERIAKKLKKDTSANVKSAGEGEVEKKSVSRSEELRKEARQLKRELLAAKQKKVENAAKQAEKRSEEEEAPPDGAVAEYRREKQKYEALRKQQSKKGTSREDQDVTCTSV
Specifications

Specifications

AAH12117.1
Liquid
10283
SDCCAG10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MANAGSHDNGSQFFFTLGRADELNNKHTIFGKVTGDTVYNMLRLSEVDIDDDERPHNPHKIKSCEVLFNPFDDIIPREIKRLKKEKPEEEVKKLKPKGTKNFSLLSFGEEAEEEEEEVNRVSQSMKGKSKSSHDLLKDDPHLSSVPVVESEKGDAPDLVDDGEDESAEHDEYIDGDEKNLMRERIAKKLKKDTSANVKSAGEGEVEKKSVSRSEELRKEARQLKRELLAAKQKKVENAAKQAEKRSEEEEAPPDGAVAEYRREKQKYEALRKQQSKKGTSREDQDVTCTSV
RUO
SDCCAG10
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NY-CO-10
SDCCAG10
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers