Learn More
Abnova™ Human SDPR Full-length ORF (NP_004648.1, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008436-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a calcium-independent phospholipid-binding protein whose expression increases in serum-starved cells. This protein is a substrate for protein kinase C (PKC) phosphorylation and recruits polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation
Sequence: MGEDAAQAEKFQHPGSDMRQEKPSSPSPMPSSTPSPSLNLGNTEEAIRDNSQVNAVTVLTLLDKLVNMLDAVQENQHKMEQRQISLEGSVKGIQNDLTKLSKYQASTSNTVSKLLEKSRKVSAHTRAVKERMDRQCAQVKRLENNHAQLLRRNHFKVLIFQEENEIPASVFVKQPVSGAVEGKEELPDENKSLEETLHTVDLSSDDDLPHDEEALEDSAEEKVEESRAEKIKRSSLKKVDSLKKAFSRQNIEKKMNKLGTKIVSVERREKIKKSLTSNHQKISSGKSSPFKVSPLTFGRKKVREGESHAENETKSEDLPSSEQMPNDQEEESFAEGHSEASLASALVEGEIAEEAAEKATSRGSNSGMDSNIDLTIVEDEEEESVALEQAQKVRYEGSYALTSEEAERSDGDPVQPAVLQVHQTSSpecifications
NP_004648.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PS-p68/SDR | |
SDPR | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8436 | |
SDPR (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGEDAAQAEKFQHPGSDMRQEKPSSPSPMPSSTPSPSLNLGNTEEAIRDNSQVNAVTVLTLLDKLVNMLDAVQENQHKMEQRQISLEGSVKGIQNDLTKLSKYQASTSNTVSKLLEKSRKVSAHTRAVKERMDRQCAQVKRLENNHAQLLRRNHFKVLIFQEENEIPASVFVKQPVSGAVEGKEELPDENKSLEETLHTVDLSSDDDLPHDEEALEDSAEEKVEESRAEKIKRSSLKKVDSLKKAFSRQNIEKKMNKLGTKIVSVERREKIKKSLTSNHQKISSGKSSPFKVSPLTFGRKKVREGESHAENETKSEDLPSSEQMPNDQEEESFAEGHSEASLASALVEGEIAEEAAEKATSRGSNSGMDSNIDLTIVEDEEEESVALEQAQKVRYEGSYALTSEEAERSDGDPVQPAVLQVHQTS | |
RUO | |
SDPR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |