Learn More
Abnova™ Human SEC22C Full-length ORF (NP_116752.1, 1 a.a. - 303 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009117-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the SEC22 family of vesicle trafficking proteins. It is localized at the endoplasmic reticulum and it is thought to play a role in the early stages of the ER-Golgi protein trafficking. There are two alternatively spliced transcript variants encoding different isoforms described for this gene. [provided by RefSeq]
Sequence: MSVIFFACVVRVRDGLPLSASTDFYHTQDFLEWRRRLKSLALRLAQYPGRGSAEGCDFSIHFSSFGDVACMAICSCQCPAAMAFCFLETLWWEFTASYDTTCIGLASRPYAFLEFDSIIQKVKWHFNYVSSSQMECSLEKIQEELKLQPPAVLTLEDTDVANGVMNGHTPMHLEPAPNFRMEPVTALGILSLILNIMCAALNLIRGVHLAEHSLQVAHEEIGNILAFLVPFVACIFQCYLYLFYSPARTMKVVLMLLFICLGNMYLHGLRNLWQILFHIGVAFLSSYQILTRQLQEKQSDCGVSpecifications
NP_116752.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761F2321/MGC13261/MGC5373/SEC22L3 | |
SEC22C | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9117 | |
SEC22C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVIFFACVVRVRDGLPLSASTDFYHTQDFLEWRRRLKSLALRLAQYPGRGSAEGCDFSIHFSSFGDVACMAICSCQCPAAMAFCFLETLWWEFTASYDTTCIGLASRPYAFLEFDSIIQKVKWHFNYVSSSQMECSLEKIQEELKLQPPAVLTLEDTDVANGVMNGHTPMHLEPAPNFRMEPVTALGILSLILNIMCAALNLIRGVHLAEHSLQVAHEEIGNILAFLVPFVACIFQCYLYLFYSPARTMKVVLMLLFICLGNMYLHGLRNLWQILFHIGVAFLSSYQILTRQLQEKQSDCGV | |
RUO | |
SEC22C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |