missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEC24A Partial ORF (XP_094581.5, 301 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | XP_094581.5 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10802 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173216
|
Abnova™
H00010802-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16163216
|
Abnova™
H00010802-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
In yeast, the Sec23-Sec24 complex is a component of coat protein II (COPII; see MIM 601924)-coated vesicles that mediate protein transport from the endoplasmic reticulum. SEC24A is 1 of several mammalian proteins that show structural and functional homology to yeast Sec24.[supplied by OMIM]
Sequence: PLTSSYRDVPQPLFNSAVNQEGITSNTNNGSMVVHSSYDEIEGGGLLATPQLTNKNPKMSRSVGYSYPSLPPGYQNTTPPGATGVPPSSLSpecifications
XP_094581.5 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SEC24A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10802 | |
SEC24A (Human) Recombinant Protein (Q01) | |
PLTSSYRDVPQPLFNSAVNQEGITSNTNNGSMVVHSSYDEIEGGGLLATPQLTNKNPKMSRSVGYSYPSLPPGYQNTTPPGATGVPPSSL | |
RUO | |
SEC24A | |
Recombinant | |
wheat germ expression system |