missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEMA4D Partial ORF (AAH54500, 115 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010507-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: LQPLSATSLYVCGTNAFQPACDHLNLTSFKFLGKNEDGKGRCPFDPAHSYTSVMVDGELYSGTSYNFLGSEPIISRNSSHSPLRTEYAIPWLNEPSFVFADVIRKSPDSPSpecifications
AAH54500 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LQPLSATSLYVCGTNAFQPACDHLNLTSFKFLGKNEDGKGRCPFDPAHSYTSVMVDGELYSGTSYNFLGSEPIISRNSSHSPLRTEYAIPWLNEPSFVFADVIRKSPDSP | |
RUO | |
SEMA4D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10507 | |
SEMA4D (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C9orf164/CD100/FLJ33485/FLJ34282/FLJ39737/FLJ46484/M-sema-G/MGC169138/MGC169141/SEMAJ/coll-4 | |
SEMA4D | |
Recombinant | |
wheat germ expression system |